Assay Details
Target Gene Details
Entrez Gene ID: | 55872 |
Gene Name: | PDZ binding kinase |
Gene Aliases: |
CT84, HEL164, Nori-3, SPK, TOPK |
Location: |
Chr.8:27809621-27838095 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PBK | NM_001278945.1 | NP_001265874.1 | ||
| NM_018492.3 | NP_060962.2 | |||
| XM_006716368.2 | XP_006716431.1 | |||
| AB027249.1 | BAA99576.1 | |||
| AB027250.1 | BAA99577.1 | |||
| AF189722.1 | AAF69107.1 | |||
| AF237709.1 | AAF71521.1 | |||
| AK027291.1 | BAB55019.1 | |||
| AK301836.1 | ||||
| BC015191.1 | AAH15191.1 | |||
| BI461026.1 | ||||
| DB028500.1 | ||||
| EU794685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524133 | Chr.8:27776547 - 27858791 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map