Assay Details
Target Gene Details
Entrez Gene ID: | 5058 |
Gene Name: | p21 (RAC1) activated kinase 1 |
Gene Aliases: |
PAKalpha |
Location: |
Chr.11:77322015-77474063 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAK1 | NM_001128620.1 | NP_001122092.1 | ||
| NM_002576.4 | NP_002567.3 | |||
| XM_011545080.2 | XP_011543382.1 | |||
| XM_011545081.2 | XP_011543383.1 | |||
| XM_011545082.2 | XP_011543384.1 | |||
| XM_011545083.2 | XP_011543385.1 | |||
| XM_011545084.2 | XP_011543386.1 | |||
| XM_017017846.1 | XP_016873335.1 | |||
| XM_017017847.1 | XP_016873336.1 | |||
| XM_017017848.1 | XP_016873337.1 | |||
| XM_017017849.1 | XP_016873338.1 | |||
| XM_017017850.1 | XP_016873339.1 | |||
| AF071884.1 | AAC24716.1 | |||
| AK055228.1 | ||||
| AK293098.1 | ||||
| AK295344.1 | ||||
| AK296006.1 | ||||
| BC109299.1 | AAI09300.1 | |||
| U24152.1 | AAA65441.1 | |||
| U51120.1 | AAC50590.1 |
More Information
Set Membership: |
|

Genomic Map