Assay Details
Target Gene Details
Entrez Gene ID: | 54979 |
Gene Name: | HRAS like suppressor 2 |
Gene Aliases: |
PLA1/2-2 |
Location: |
Chr.11:63552759-63565072 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HRASLS2 | NM_017878.1 | NP_060348.1 | ||
XM_011545120.2 | XP_011543422.1 | |||
AK000563.1 | BAA91256.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3626647 | Chr.11:63558459 - 63582290 on Build GRCh38 | Gain |
![]() ![]() |
esv3626646 | Chr.11:63549774 - 63585192 on Build GRCh38 | Gain |
![]() ![]() |
nsv352 | Chr.11:63555836 - 63589650 on Build GRCh38 | Insertion |
![]() ![]() |
nsv509417 | Chr.11:63485643 - 63569743 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
