Assay Details
Target Gene Details
Entrez Gene ID: | 196264 |
Gene Name: | myelin protein zero like 3 |
Gene Aliases: |
- |
Location: |
Chr.11:118226690-118252368 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPZL3 | NM_001286152.1 | 6 | 2877 | NP_001273081.1 |
| NM_198275.2 | 6 | 2913 | NP_938016.1 | |
| NR_104404.1 | 3 | 2369 | ||
| NR_104405.1 | 5 | 2702 | ||
| XM_006718775.2 | 6 | 3265 | XP_006718838.1 | |
| XM_017017304.1 | 6 | 3024 | XP_016872793.1 | |
| AK095399.1 | 6 | 2865 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv511 | Chr.11:118204437 - 118239426 on Build GRCh38 | Insertion |
|
| nsv952742 | Chr.11:118218486 - 118228985 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map