Assay Details
Target Gene Details
Entrez Gene ID: | 1717 |
Gene Name: | 7-dehydrocholesterol reductase |
Gene Aliases: |
SLOS |
Location: |
Chr.11:71434411-71448431 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHCR7 | NM_001163817.1 | NP_001157289.1 | ||
| NM_001360.2 | NP_001351.2 | |||
| XM_011544777.1 | XP_011543079.1 | |||
| AF034544.1 | AAC05086.1 | |||
| AF062481.1 | AAC18345.1 | |||
| AF067127.1 | AAD02816.1 | |||
| AF096305.1 | AAD09766.1 | |||
| AK289497.1 | ||||
| AK303881.1 | ||||
| AK309625.1 | ||||
| AK312775.1 | ||||
| BC000054.2 | AAH00054.1 | |||
| KJ534821.1 | ||||
| KJ534963.1 | ||||
| KJ534997.1 | ||||
| KU178026.1 | ||||
| KU178027.1 | ||||
| KU178028.1 | ||||
| KU178029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv74n21 | Chr.11:71357749 - 71504834 on Build GRCh38 | Loss |
|
| nsv518163 | Chr.11:71431304 - 71444107 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map