Assay Details
Target Gene Details
Entrez Gene ID: | 11309 |
Gene Name: | solute carrier organic anion transporter family member 2B1 |
Gene Aliases: |
OATP-B, OATP2B1, OATPB, SLC21A9 |
Location: |
Chr.11:75150987-75206400 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLCO2B1 | NM_001145211.2 | NP_001138683.1 | ||
| NM_001145212.2 | NP_001138684.1 | |||
| NM_007256.4 | NP_009187.1 | |||
| XM_017017157.1 | XP_016872646.1 | |||
| AB020687.1 | ||||
| AB026256.1 | BAA78638.1 | |||
| AF205073.1 | AAG42205.1 | |||
| AK290234.1 | ||||
| AK294503.1 | ||||
| AK300134.1 | ||||
| AK302492.1 | ||||
| AK304783.1 | ||||
| AL117465.1 | CAB55940.1 | |||
| BC041095.1 | AAH41095.1 | |||
| DC335261.1 | ||||
| DC343392.1 | ||||
| DC362722.1 | ||||
| FM209049.1 | ||||
| FM209050.1 | ||||
| FM209051.1 | ||||
| FM209052.1 | ||||
| FM209053.1 | ||||
| FM209054.1 |
More Information
Set Membership: |
|

Genomic Map