Assay Details
Target Gene Details
Entrez Gene ID: | 105376671 |
Gene Name: | uncharacterized LOC105376671 |
Gene Aliases: |
lnc-BBOX1-1 |
Location: |
Chr.11:27471731-27482433 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105376671 | NR_131169.1 | 3 | 513 | |
| NR_131170.1 | ||||
| AF150379.1 | 3 | 514 | ||
| DA085906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1070018 | Chr.11:27470552 - 27473553 on Build GRCh38 | Deletion |
|
| nsv467782 | Chr.11:27171168 - 27474614 on Build GRCh38 | Gain |
|

Genomic Map