Assay Details
Target Gene Details
Entrez Gene ID: | 53340 |
Gene Name: | sperm autoantigenic protein 17 |
Gene Aliases: |
CT22, SP17, SP17-1 |
Location: |
Chr.11:124673844-124694794 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPA17 | NM_017425.3 | NP_059121.1 | ||
| XM_011542870.2 | XP_011541172.1 | |||
| AF334735.1 | AAK20878.1 | |||
| AK307271.1 | ||||
| AK311806.1 | ||||
| BC019224.1 | ||||
| BC032457.2 | AAH32457.1 | |||
| DA511916.1 | ||||
| KU257683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045156 | Chr.11:124565356 - 124705852 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map