Assay Details
Target Gene Details
Entrez Gene ID: | 10418 |
Gene Name: | spondin 1 |
Gene Aliases: |
VSGP/F-spondin, f-spondin |
Location: |
Chr.11:13962637-14268133 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPON1 | NM_006108.3 | NP_006099.2 | ||
| AB051390.1 | BAB18461.1 | |||
| AK074803.1 | BAC11217.1 | |||
| AK291780.1 | ||||
| BC136513.1 | ||||
| BC136563.1 | ||||
| DA965056.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951310 | Chr.11:13961154 - 13964653 on Build GRCh38 | Deletion |
|
| esv2668036 | Chr.11:13962055 - 14027138 on Build GRCh38 | Deletion |
|
| esv3625418 | Chr.11:13962063 - 14027140 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map