Assay Details
Target Gene Details
Entrez Gene ID: | 65975 |
Gene Name: | serine/threonine kinase 33 |
Gene Aliases: |
- |
Location: |
Chr.11:8335029-8594289 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK33 |
More Information
Additional Information:
For this assay, SNP(s) [rs73400100,rs77141343] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map