Assay Details
Target Gene Details
Entrez Gene ID: | 22992 |
Gene Name: | lysine demethylase 2A |
Gene Aliases: |
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA |
Location: |
Chr.11:67119210-67258082 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM2A | NM_012308.2 | NP_036440.1 | ||
| NR_027473.1 | ||||
| XM_011544860.2 | 1 | 58 | XP_011543162.1 | |
| BC047486.1 | AAH47486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1069600 | Chr.11:67118028 - 67119529 on Build GRCh38 | Deletion |
|
| nsv1121839 | Chr.11:67113829 - 67119729 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map