Assay Details
Target Gene Details
Entrez Gene ID: | 79080 |
Gene Name: | coiled-coil domain containing 86 |
Gene Aliases: |
- |
Location: |
Chr.11:60841956-60851088 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC86 | NM_024098.3 | NP_077003.1 | ||
| AK025974.1 | BAB15304.1 | |||
| AK123775.1 | BAC85689.1 | |||
| AK302329.1 | ||||
| BC001378.1 | AAH01378.1 | |||
| DQ501251.1 |
More Information
Set Membership: |
|

Genomic Map