Assay Details
Target Gene Details
Entrez Gene ID: | 10196 |
Gene Name: | protein arginine methyltransferase 3 |
Gene Aliases: |
HRMT1L3 |
Location: |
Chr.11:20387530-20510861 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRMT3 | NM_001145166.1 | NP_001138638.1 | ||
| NM_001145167.1 | NP_001138639.1 | |||
| NM_005788.3 | NP_005779.1 | |||
| XM_011519836.2 | XP_011518138.1 | |||
| XM_017017081.1 | XP_016872570.1 | |||
| AF059531.1 | AAC39837.1 | |||
| AK129984.1 | ||||
| AK298185.1 | ||||
| AK300591.1 | ||||
| AK301449.1 | ||||
| AK307776.1 | ||||
| BC019339.1 | AAH19339.1 | |||
| BC037544.1 | AAH37544.1 | |||
| BC064831.1 | AAH64831.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553678 | Chr.11:20097752 - 20718575 on Build GRCh38 | Gain |
|
| nsv832083 | Chr.11:20318953 - 20490151 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map