Assay Details
Target Gene Details
Entrez Gene ID: | 50863 |
Gene Name: | neurotrimin |
Gene Aliases: |
HNT, IGLON2, NTRI |
Location: |
Chr.11:131370326-132336822 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Gene Details
Entrez Gene ID: | 100874281 |
Gene Name: | NTM intronic transcript |
Gene Aliases: |
NTM-IT3 |
Location: |
Chr.11:132284302-132288459 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTM-IT | NR_046820.1 | |||
| AA435898.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892111 | Chr.11:132278058 - 132297744 on Build GRCh38 | Loss |
|
| esv3628121 | Chr.11:132280887 - 132292976 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map