Assay Details
Target Gene Details
Entrez Gene ID: | 26973 |
Gene Name: | cysteine and histidine rich domain containing 1 |
Gene Aliases: |
CHP1 |
Location: |
Chr.11:90200429-90223364 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHORDC1 | NM_001144073.1 | NP_001137545.1 | ||
| NM_012124.2 | NP_036256.2 | |||
| XM_017017540.1 | XP_016873029.1 | |||
| XM_017017541.1 | XP_016873030.1 | |||
| AF123249.1 | AAG43237.1 | |||
| AF192466.1 | AAF18437.1 | |||
| AK225365.1 | ||||
| AK225565.1 | ||||
| AK226099.1 | ||||
| AK290231.1 | ||||
| AK312663.1 | ||||
| BC072461.1 | AAH72461.1 | |||
| DA417105.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555833 | Chr.11:90162772 - 90275439 on Build GRCh38 | Loss |
|
| esv3579993 | Chr.11:89580840 - 90609205 on Build GRCh38 | Gain |
|
| esv3579841 | Chr.11:90080153 - 90413922 on Build GRCh38 | Loss |
|
| nsv1035213 | Chr.11:90152198 - 90257741 on Build GRCh38 | Gain |
|
| nsv1048624 | Chr.11:90100568 - 91604298 on Build GRCh38 | Loss |
|
| nsv468787 | Chr.11:90168986 - 90220213 on Build GRCh38 | Loss |
|
| nsv555832 | Chr.11:90142496 - 90309371 on Build GRCh38 | Gain |
|
| nsv1048042 | Chr.11:90191353 - 90229342 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77975598] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map