Assay Details
Target Gene Details
Entrez Gene ID: | 9479 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 1 |
Gene Aliases: |
IB1, JIP-1, JIP1, PRKM8IP |
Location: |
Chr.11:45885496-45906465 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK8IP1 | NM_005456.3 | NP_005447.1 | ||
| AF074091.1 | AAD20443.1 | |||
| BC068470.1 | AAH68470.1 | |||
| BI548334.1 | ||||
| DB473314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516442 | Chr.11:45878411 - 45961834 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map