Assay Details
Target Gene Details
Entrez Gene ID: | 101928896 |
Gene Name: | uncharacterized LOC101928896 |
Gene Aliases: |
- |
Location: |
Chr.11:78533176-78558565 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928896 | NR_120566.1 |
Target Gene Details
Entrez Gene ID: | 79731 |
Gene Name: | asparaginyl-tRNA synthetase 2, mitochondrial (putative) |
Gene Aliases: |
DFNB94, SLM5, asnRS |
Location: |
Chr.11:78435961-78574864 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NARS2 | NM_001243251.1 | NP_001230180.1 | ||
| NM_024678.5 | NP_078954.4 | |||
| XM_011545253.1 | XP_011543555.1 | |||
| XM_017018302.1 | XP_016873791.1 | |||
| XM_017018303.1 | XP_016873792.1 | |||
| XM_017018304.1 | XP_016873793.1 | |||
| AK027094.1 | BAB15655.1 | |||
| AK056980.1 | ||||
| AK225444.1 | ||||
| AK315425.1 | ||||
| BC007800.2 | AAH07800.2 | |||
| BP196275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv211e214 | Chr.11:78524451 - 78661281 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map