Assay Details
Target Gene Details
Entrez Gene ID: | 55742 |
Gene Name: | parvin alpha |
Gene Aliases: |
CH-ILKBP, MXRA2 |
Location: |
Chr.11:12376441-12535356 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARVA | NM_018222.4 | NP_060692.2 | ||
| XM_005253015.3 | XP_005253072.1 | |||
| AF237771.1 | AAG27173.1 | |||
| AF325830.1 | AAK49911.1 | |||
| AK001655.1 | BAA91815.1 | |||
| AK022316.1 | BAB14009.1 | |||
| AK098504.1 | ||||
| BC014535.1 | AAH14535.1 | |||
| BC016713.1 | AAH16713.1 | |||
| DC344538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625390 | Chr.11:12320996 - 12405629 on Build GRCh38 | Gain |
|
| esv3625391 | Chr.11:12333021 - 12387704 on Build GRCh38 | Gain |
|
| nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss |
|
| dgv1059n100 | Chr.11:12197656 - 12685051 on Build GRCh38 | Gain |
|
| esv3579371 | Chr.11:12372526 - 12379600 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map