Assay Details
Target Gene Details
Entrez Gene ID: | 5893 |
Gene Name: | RAD52 homolog, DNA repair protein |
Gene Aliases: |
- |
Location: |
Chr.12:911028-991195 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain |
|
| esv3628261 | Chr.12:880274 - 938296 on Build GRCh38 | Gain |
|
| nsv1042333 | Chr.12:798809 - 972343 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map