Assay Details
Target Gene Details
Entrez Gene ID: | 4908 |
Gene Name: | neurotrophin 3 |
Gene Aliases: |
HDNF, NGF-2, NGF2, NT-3, NT3 |
Location: |
Chr.12:5432114-5495299 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTF3 | NM_001102654.1 | NP_001096124.1 | ||
| XM_011520963.2 | XP_011519265.1 | |||
| AK293895.1 | ||||
| BU939830.1 | ||||
| CN267386.1 | ||||
| X53655.1 | CAA37703.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468988 | Chr.12:5413390 - 6141521 on Build GRCh38 | Gain |
|
| nsv592 | Chr.12:5440123 - 5444838 on Build GRCh38 | Insertion |
|
| nsv1040098 | Chr.12:3987637 - 5646161 on Build GRCh38 | Gain |
|
| nsv523152 | Chr.12:5428946 - 5588893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map