Assay Details
Target Gene Details
Entrez Gene ID: | 160428 |
Gene Name: | aldehyde dehydrogenase 1 family member L2 |
Gene Aliases: |
mtFDH |
Location: |
Chr.12:105019784-105084609 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH1L2 | NM_001034173.3 | 23 | 7153 | NP_001029345.2 |
| NR_027752.1 | 23 | 7105 | ||
| XM_011537986.2 | 21 | 6712 | XP_011536288.1 | |
| XM_017018889.1 | 24 | 7359 | XP_016874378.1 | |
| AK094088.1 | 1 | 1327 | ||
| BX537599.1 | 1 | 3517 | ||
| CA418450.1 | 1 | 403 | ||
| CR627287.1 | 19 | 7017 | CAH10368.1 | |
| CR749561.1 | 4 | 4745 | CAH18358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832505 | Chr.12:104921574 - 105103987 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs147185195] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map