Assay Details
Target Gene Details
Entrez Gene ID: | 121665 |
Gene Name: | signal peptide peptidase like 3 |
Gene Aliases: |
IMP2, MDHV1887, PRO4332, PSH1, PSL4 |
Location: |
Chr.12:120762510-120904352 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPPL3 | NM_139015.4 | NP_620584.2 | ||
| XM_011537925.2 | XP_011536227.1 | |||
| AB252457.1 | ||||
| AJ345030.1 | CAC87791.1 | |||
| AJ420898.1 | CAD13135.1 | |||
| AK074916.1 | BAC11290.1 | |||
| AK097149.1 | 1 | 1692 | ||
| AK223590.1 | BAD97310.1 | |||
| AK291161.1 | ||||
| AY169313.1 | AAO12538.1 | |||
| BC025781.1 | AAH25781.1 | |||
| BC073910.1 | AAH73910.1 | |||
| BC101625.1 | AAI01626.1 | |||
| BC101627.1 | AAI01628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
|
| nsv560438 | Chr.12:120756762 - 120914403 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map