Assay Details
Target Gene Details
Entrez Gene ID: | 517 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9) |
Gene Aliases: |
ATP5A |
Location: |
Chr.12:53665160-53677546 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5G2 | NM_001002031.2 | NP_001002031.1 | ||
| NM_005176.5 | NP_005167.2 | |||
| XM_017019460.1 | XP_016874949.1 | |||
| XM_017019461.1 | XP_016874950.1 | |||
| XM_017019462.1 | XP_016874951.1 | |||
| AK075351.1 | 1 | 584 | ||
| AK130971.1 | ||||
| AK311357.1 | ||||
| BF337574.1 | ||||
| BG217012.1 | ||||
| BX348847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526791 | Chr.12:53561701 - 53700252 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map