Assay Details
Target Gene Details
Entrez Gene ID: | 9498 |
Gene Name: | solute carrier family 4 member 8 |
Gene Aliases: |
NBC3, NDCBE |
Location: |
Chr.12:51391317-51515763 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A8 | NM_001258401.2 | NP_001245330.1 | ||
| NM_001267615.1 | NP_001254544.1 | |||
| XM_011539011.1 | XP_011537313.1 | |||
| XM_017020241.1 | 1 | 200 | XP_016875730.1 | |
| AB018282.1 | BAA34459.1 | |||
| AK128321.1 | ||||
| AK295315.1 | ||||
| DQ975204.1 | ||||
| DQ996398.1 | ||||
| DQ996537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745884 | Chr.12:51402880 - 51403457 on Build GRCh38 | Deletion |
|
| nsv1045796 | Chr.12:51400259 - 51450243 on Build GRCh38 | Loss |
|
| esv3629502 | Chr.12:51399705 - 51409756 on Build GRCh38 | Gain |
|
| nsv832408 | Chr.12:51302821 - 51477555 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map