Assay Details
Target Gene Details
Entrez Gene ID: | 387836 |
Gene Name: | C-type lectin domain family 2 member A |
Gene Aliases: |
INPE5792, KACL, PILAR, UNQ5792 |
Location: |
Chr.12:9882733-9932430 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC2A | XM_011520657.2 | XP_011518959.1 |
Target Gene Details
Entrez Gene ID: | 100431172 |
Gene Name: | killer cell lectin like receptor F2 |
Gene Aliases: |
NKp65 |
Location: |
Chr.12:9881489-9895833 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLRF2 | NM_001190765.1 | NP_001177694.1 | ||
| GQ398770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv1044587 | Chr.12:9876915 - 9970815 on Build GRCh38 | Loss |
|
| nsv523050 | Chr.12:9879281 - 9971774 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map