Assay Details
Target Gene Details
Entrez Gene ID: | 55729 |
Gene Name: | activating transcription factor 7 interacting protein |
Gene Aliases: |
AM, ATF-IP, MCAF, MCAF1, p621 |
Location: |
Chr.12:14365632-14502935 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF7IP | NM_001286514.1 | NP_001273443.1 | ||
| NM_001286515.1 | NP_001273444.1 | |||
| NM_018179.4 | NP_060649.3 | |||
| NM_181352.1 | NP_851997.1 | |||
| XM_005253424.3 | XP_005253481.1 | |||
| XM_011520755.2 | XP_011519057.1 | |||
| XM_011520756.2 | XP_011519058.1 | |||
| XM_017019638.1 | XP_016875127.1 | |||
| AK025060.1 | ||||
| AK299320.1 | ||||
| AK304184.1 | ||||
| AK308062.1 | ||||
| BC063855.1 | AAH63855.1 | |||
| BX648096.1 | ||||
| DA194386.1 | ||||
| DC377662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1393n100 | Chr.12:14156211 - 14470137 on Build GRCh38 | Gain |
|
| nsv1047428 | Chr.12:14398551 - 14456148 on Build GRCh38 | Loss |
|
| nsv951332 | Chr.12:14389867 - 14466966 on Build GRCh38 | Duplication |
|
| nsv832335 | Chr.12:14255954 - 14453814 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map