Assay Details
Target Gene Details
Entrez Gene ID: | 204010 |
Gene Name: | ribosomal protein SA pseudogene 52 |
Gene Aliases: |
RPSA_17_1251 |
Location: |
Chr.12:65758020-65826974 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPSAP52 | NR_026825.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs138748438] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map