Assay Details
Target Gene Details
Entrez Gene ID: | 8835 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2 |
Location: |
Chr.12:93569822-93626236 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
More Information
Set Membership: |
|

Genomic Map