Assay Details
Target Gene Details
Entrez Gene ID: | 3479 |
Gene Name: | insulin like growth factor 1 |
Gene Aliases: |
IGF-I, IGFI, MGF |
Location: |
Chr.12:102395867-102481839 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGF1 | NM_000618.4 | NP_000609.1 | ||
| NM_001111283.2 | NP_001104753.1 | |||
| NM_001111284.1 | NP_001104754.1 | |||
| XM_017019262.1 | XP_016874751.1 | |||
| XM_017019263.1 | XP_016874752.1 | |||
| AB209184.1 | BAD92421.1 | |||
| AK312231.1 | ||||
| BC152321.1 | ||||
| CR541861.1 | CAG46659.1 | |||
| M27544.1 | AAA52787.1 | |||
| M29644.1 | AAA52543.1 | |||
| M37484.1 | AAA52789.1 | |||
| U40870.1 | AAA96152.1 | |||
| X00173.1 | CAA24998.1 | |||
| X56773.1 | CAA40092.1 | |||
| X57025.1 | CAA40342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832498 | Chr.12:102334798 - 102519791 on Build GRCh38 | Gain |
|
| nsv560035 | Chr.12:102407157 - 102413892 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map