Assay Details
Target Gene Details
Entrez Gene ID: | 54934 |
Gene Name: | KAT8 regulatory NSL complex subunit 2 |
Gene Aliases: |
C12orf41, NSL2 |
Location: |
Chr.12:48653211-48682252 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANSL2 | NM_017822.3 | NP_060292.3 | ||
| AK000443.1 | BAA91169.1 | |||
| AK022732.1 | BAB14211.1 | |||
| AK094528.1 | ||||
| AL834467.1 | CAD39126.1 | |||
| BC009746.2 | AAH09746.1 | |||
| BC013900.2 | AAH13900.1 | |||
| BG760446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1496n100 | Chr.12:48651675 - 48672204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map