Assay Details
Target Gene Details
Entrez Gene ID: | 5213 |
Gene Name: | phosphofructokinase, muscle |
Gene Aliases: |
ATP-PFK, GSD7, PFK-1, PFK1, PFKA, PFKX, PPP1R122 |
Location: |
Chr.12:48105278-48146404 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PFKM | NM_001166686.1 | NP_001160158.1 | ||
| XM_005268974.1 | XP_005269031.1 | |||
| XM_005268975.1 | XP_005269032.1 | |||
| XM_005268976.3 | XP_005269033.1 | |||
| XM_005268977.1 | XP_005269034.1 | |||
| XM_005268978.2 | XP_005269035.1 | |||
| XM_011538488.2 | XP_011536790.1 | |||
| XM_017019467.1 | XP_016874956.1 | |||
| XM_017019468.1 | XP_016874957.1 | |||
| XM_017019469.1 | XP_016874958.1 | |||
| AK126020.1 | ||||
| AK126229.1 | BAC86498.1 | |||
| DB081387.1 |
Target Gene Details
Entrez Gene ID: | 29843 |
Gene Name: | SUMO1/sentrin specific peptidase 1 |
Gene Aliases: |
SuPr-2 |
Location: |
Chr.12:48042893-48106308 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP1 | NM_001267594.1 | NP_001254523.1 | ||
| XM_017019237.1 | XP_016874726.1 | |||
| AK312874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759898 | Chr.12:48035919 - 48431716 on Build GRCh38 | Gain+Loss |
|
| nsv508673 | Chr.12:48096003 - 48173355 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map