Assay Details
Target Gene Details
Entrez Gene ID: | 79912 |
Gene Name: | pyridine nucleotide-disulphide oxidoreductase domain 1 |
Gene Aliases: |
- |
Location: |
Chr.12:21437604-21471250 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYROXD1 | NM_024854.3 | NP_079130.2 | ||
| XM_017019976.1 | XP_016875465.1 | |||
| AK025681.1 | BAB15214.1 | |||
| AK125461.1 | ||||
| AK307710.1 | ||||
| AL832441.1 | CAH10632.1 | |||
| BC021662.1 | AAH21662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557741 | Chr.12:21425625 - 22016920 on Build GRCh38 | Gain |
|
| nsv1054220 | Chr.12:21459224 - 21539169 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map