Assay Details
Target Gene Details
Entrez Gene ID: | 196446 |
Gene Name: | myelin regulatory factor-like |
Gene Aliases: |
C12orf15, C12orf28, bcm1377 |
Location: |
Chr.12:69824518-69958726 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYRFL | NM_182530.2 | NP_872336.2 | ||
| XM_011539052.2 | XP_011537354.1 | |||
| XM_017018968.1 | XP_016874457.1 | |||
| XM_017018971.1 | XP_016874460.1 | |||
| XM_017018972.1 | XP_016874461.1 | |||
| XM_017018974.1 | XP_016874463.1 | |||
| XM_017018975.1 | XP_016874464.1 | |||
| XM_017018976.1 | XP_016874465.1 |
More Information
Set Membership: |
|

Genomic Map