Assay Details
Target Gene Details
Entrez Gene ID: | 2841 |
Gene Name: | G protein-coupled receptor 18 |
Gene Aliases: |
- |
Location: |
Chr.13:99254713-99262498 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR18 | XM_006719946.3 | XP_006720009.1 | ||
| BC066927.1 | AAH66927.1 |
Target Gene Details
Entrez Gene ID: | 337867 |
Gene Name: | UBA domain containing 2 |
Gene Aliases: |
PHGDHL1 |
Location: |
Chr.13:99200425-99386499 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBAC2 | NM_001144072.1 | NP_001137544.1 | ||
| NM_177967.3 | NP_808882.1 | |||
| NR_026644.1 | ||||
| XM_006719948.3 | XP_006720011.1 | |||
| XM_011521082.2 | XP_011519384.1 | |||
| XM_011521083.2 | XP_011519385.1 | |||
| XM_011521084.2 | XP_011519386.1 | |||
| AK054563.1 | BAB70757.1 | |||
| AK075419.1 | BAC11609.1 | |||
| AK075516.1 | BAC11665.1 | |||
| AK124404.1 | ||||
| AK129861.1 | ||||
| AK290342.1 | ||||
| AK300908.1 | ||||
| BC063559.1 | AAH63559.1 | |||
| BC121138.1 | ||||
| BC121139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050094 | Chr.13:98974062 - 99323969 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs571495872] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map