Assay Details
Target Gene Details
Entrez Gene ID: | 83852 |
Gene Name: | SET domain bifurcated 2 |
Gene Aliases: |
C13orf4, CLLD8, CLLL8, KMT1F |
Location: |
Chr.13:49444293-49495003 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETDB2 | NM_001160308.2 | NP_001153780.1 | ||
| NM_001320699.1 | NP_001307628.1 | |||
| NM_031915.2 | NP_114121.2 | |||
| AF334407.1 | AAK38373.1 | |||
| AK307593.1 | ||||
| AK308220.1 | ||||
| AK308354.1 | ||||
| BC047434.2 | AAH47434.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561613 | Chr.13:49439486 - 49512130 on Build GRCh38 | Loss |
|
| nsv523648 | Chr.13:49431308 - 49512130 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map