Assay Details
Target Gene Details
Entrez Gene ID: | 688 |
Gene Name: | Kruppel like factor 5 |
Gene Aliases: |
BTEB2, CKLF, IKLF |
Location: |
Chr.13:73054976-73077542 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF5 | NM_001286818.1 | NP_001273747.1 | ||
| NM_001730.4 | NP_001721.2 | |||
| AB030824.1 | BAA96461.1 | |||
| AF132818.1 | AAF18307.1 | |||
| AF287272.1 | AAF88068.1 | |||
| AK131397.1 | 1 | 561 | ||
| AK296900.1 | ||||
| AK302280.1 | ||||
| AK304284.1 | ||||
| BC007695.1 | AAH07695.1 | |||
| BC042131.1 | AAH42131.1 | |||
| BT006641.1 | AAP35287.1 | |||
| D14520.1 | BAA03393.1 | |||
| EF208215.1 | ||||
| HF546203.1 | ||||
| HF546204.1 | ||||
| HQ628639.1 | ||||
| HQ628640.1 | ||||
| HQ628641.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78854176] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map