Assay Details
Target Gene Details
Entrez Gene ID: | 115825 |
Gene Name: | WD repeat and FYVE domain containing 2 |
Gene Aliases: |
PROF, WDF2, ZFYVE22 |
Location: |
Chr.13:51584194-51766799 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY2 | NM_052950.3 | NP_443182.1 | ||
| XM_011534914.1 | XP_011533216.1 | |||
| XM_011534915.2 | XP_011533217.1 | |||
| XM_017020386.1 | 2 | 1643 | XP_016875875.1 | |
| AF411978.1 | AAL04162.1 | |||
| AK309815.1 | ||||
| AK311177.1 | ||||
| BC014004.2 | AAH14004.1 | |||
| DA124107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892343 | Chr.13:51418581 - 52060832 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map