Assay Details
Target Gene Details
Entrez Gene ID: | 2262 |
Gene Name: | glypican 5 |
Gene Aliases: |
- |
Location: |
Chr.13:91398619-92867237 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPC5 | NM_004466.5 | NP_004457.1 | ||
| XM_011521054.2 | XP_011519356.1 | |||
| XM_011521055.2 | XP_011519357.1 | |||
| XM_011521056.2 | XP_011519358.1 | |||
| XM_011521057.2 | XP_011519359.1 | |||
| XM_011521058.2 | XP_011519360.1 | |||
| XM_011521059.2 | XP_011519361.1 | |||
| XM_011521060.2 | XP_011519362.1 | |||
| XM_017020435.1 | XP_016875924.1 | |||
| XM_017020436.1 | XP_016875925.1 | |||
| XM_017020437.1 | XP_016875926.1 | |||
| AF001462.1 | AAC12261.1 | |||
| AK312815.1 | ||||
| BC039730.1 | AAH39730.1 | |||
| DB100549.1 | ||||
| U66033.1 | AAC51118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3318n54 | Chr.13:91404657 - 91652720 on Build GRCh38 | Loss |
|
| nsv1041168 | Chr.13:91409803 - 91649607 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73614206] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map