Assay Details
Target Gene Details
Entrez Gene ID: | 3356 |
Gene Name: | 5-hydroxytryptamine receptor 2A |
Gene Aliases: |
5-HT2A, HTR2 |
Location: |
Chr.13:46831542-46897076 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR2A | NM_000621.4 | NP_000612.1 | ||
| NM_001165947.2 | NP_001159419.1 | |||
| A23337.1 | CAA01675.1 | |||
| AF498982.1 | AAM21129.1 | |||
| AK302787.1 | ||||
| AK314132.1 | ||||
| BC069356.1 | AAH69356.1 | |||
| BC069576.1 | AAH69576.1 | |||
| BC074848.2 | AAH74848.1 | |||
| BC074849.2 | AAH74849.1 | |||
| BC096839.1 | AAH96839.1 | |||
| BX647777.1 | ||||
| M86841.1 | AAA58354.1 | |||
| S71229.1 | AAB31320.1 | |||
| X57830.1 | CAA40963.1 |
Target Gene Details
Entrez Gene ID: | 100874082 |
Gene Name: | HTR2A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:46852140-46856299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR2A-AS1 | NR_046612.1 | |||
| NR_103752.1 | ||||
| AI076014.1 | ||||
| AI216351.1 | ||||
| AI914390.1 | ||||
| AW469493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763019 | Chr.13:46774094 - 47549312 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map