Assay Details
Target Gene Details
Entrez Gene ID: | 9482 |
Gene Name: | syntaxin 8 |
Gene Aliases: |
CARB |
Location: |
Chr.17:9250471-9575958 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX8 | NM_004853.2 | NP_004844.1 | ||
| NR_033656.1 | ||||
| XM_011524079.2 | XP_011522381.1 | |||
| AF036715.1 | AAC95285.1 | |||
| AF062077.1 | AAC36466.1 | |||
| AF115323.1 | AAD20831.1 | |||
| BC009713.1 | AAH09713.1 | |||
| BT007319.1 | AAP35983.1 | |||
| BU596252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064011 | Chr.17:9203289 - 9492596 on Build GRCh38 | Gain |
|
| nsv574341 | Chr.17:9148000 - 9504608 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map