Assay Details
Target Gene Details
Entrez Gene ID: | 102724508 |
Gene Name: | thyroid cancer-associated transcript 158 |
Gene Aliases: |
- |
Location: |
Chr.17:47303443-47323901 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THCAT158 | NR_110880.1 | |||
| NR_110881.1 | ||||
| AA430297.1 | ||||
| AW293117.1 | ||||
| HY036509.1 |

Genomic Map