Assay Details
Target Gene Details
Entrez Gene ID: | 1236 |
Gene Name: | C-C motif chemokine receptor 7 |
Gene Aliases: |
BLR2, CC-CKR-7, CCR-7, CD197, CDw197, CMKBR7, EBI1 |
Location: |
Chr.17:40553769-40565484 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR7 | NM_001301714.1 | NP_001288643.1 | ||
| NM_001838.3 | NP_001829.1 | |||
| AK127810.1 | ||||
| AK312732.1 | ||||
| BC035343.1 | AAH35343.1 | |||
| BI910219.1 | ||||
| DA814913.1 | ||||
| L08176.1 | AAA58615.1 | |||
| L31581.1 | AAA74231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574994 | Chr.17:40563547 - 40668384 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map