Assay Details
Target Gene Details
Entrez Gene ID: | 84152 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 1B |
Gene Aliases: |
DARPP-32, DARPP32 |
Location: |
Chr.17:39626208-39636625 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R1B | NM_032192.3 | 1 | 365 | NP_115568.2 |
| XM_017025216.1 | XP_016880705.1 | |||
| AF435972.1 | 1 | 324 | AAP35094.1 | |
| AF464196.1 | 1 | 363 | AAM76683.1 | |
| AK123112.1 | 1 | 338 | ||
| AK127542.1 | 2 | 229 | ||
| AK129537.1 | 2 | 281 | ||
| DA136306.1 | 1 | 366 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2047 | Chr.17:39602374 - 39637631 on Build GRCh38 | Insertion |
|
| nsv521294 | Chr.17:39538456 - 39648605 on Build GRCh38 | Loss |
|
| nsv457738 | Chr.17:39613752 - 39674782 on Build GRCh38 | Loss |
|
| nsv516757 | Chr.17:39625596 - 39678288 on Build GRCh38 | Loss |
|
| dgv40n68 | Chr.17:39536627 - 39726611 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map