Assay Details
Target Gene Details
Entrez Gene ID: | 10238 |
Gene Name: | DDB1 and CUL4 associated factor 7 |
Gene Aliases: |
AN11, HAN11, SWAN-1, WDR68 |
Location: |
Chr.17:63550435-63594279 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF7 | NM_005828.4 | NP_005819.3 | ||
| NR_073585.1 | ||||
| AK294925.1 | ||||
| AK303212.1 | ||||
| BQ221019.1 | ||||
| DA230140.1 | ||||
| HY023965.1 | ||||
| U94747.1 | AAC18913.1 |
More Information
Set Membership: |
|

Genomic Map