Assay Details
Target Gene Details
Entrez Gene ID: | 22806 |
Gene Name: | IKAROS family zinc finger 3 |
Gene Aliases: |
AIO, AIOLOS, ZNFN1A3 |
Location: |
Chr.17:39757715-39864188 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKZF3 | NM_001257408.1 | 7 | 9459 | NP_001244337.1 |
| NM_001257409.1 | 6 | 9342 | NP_001244338.1 | |
| NM_001257410.1 | 7 | 9300 | NP_001244339.1 | |
| NM_001257411.1 | 6 | 9132 | NP_001244340.1 | |
| NM_001257412.1 | 5 | 9015 | NP_001244341.1 | |
| NM_001257413.1 | 4 | 8898 | NP_001244342.1 | |
| NM_001257414.1 | 5 | 9159 | NP_001244343.1 | |
| NM_001284514.1 | 9 | 9687 | NP_001271443.1 | |
| NM_001284515.1 | 8 | 9519 | NP_001271444.1 | |
| NM_001284516.1 | 3 | 9131 | NP_001271445.1 | |
| NM_012481.4 | 8 | 9561 | NP_036613.2 | |
| NM_183228.2 | 7 | 9393 | NP_899051.1 | |
| NM_183229.2 | 7 | 9444 | NP_899052.1 | |
| NM_183230.2 | 7 | 9444 | NP_899053.1 | |
| NM_183231.2 | 6 | 9276 | NP_899054.1 | |
| NM_183232.2 | 6 | 9327 | NP_899055.1 | |
| AI866838.1 | 1 | 126 | ||
| AK128436.1 | 1 | 3480 |
More Information
Set Membership: |
|

Genomic Map