Assay Details
Target Gene Details
Entrez Gene ID: | 22806 |
Gene Name: | IKAROS family zinc finger 3 |
Gene Aliases: |
AIO, AIOLOS, ZNFN1A3 |
Location: |
Chr.17:39757715-39864188 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKZF3 | NM_001257408.1 | 7 | 4945 | NP_001244337.1 |
| NM_001257409.1 | 6 | 4828 | NP_001244338.1 | |
| NM_001257410.1 | 7 | 4786 | NP_001244339.1 | |
| NM_001257411.1 | 6 | 4618 | NP_001244340.1 | |
| NM_001257412.1 | 5 | 4501 | NP_001244341.1 | |
| NM_001257413.1 | 4 | 4384 | NP_001244342.1 | |
| NM_001257414.1 | 5 | 4645 | NP_001244343.1 | |
| NM_001284514.1 | 9 | 5173 | NP_001271443.1 | |
| NM_001284515.1 | 8 | 5005 | NP_001271444.1 | |
| NM_001284516.1 | 3 | 4617 | NP_001271445.1 | |
| NM_012481.4 | 8 | 5047 | NP_036613.2 | |
| NM_183228.2 | 7 | 4879 | NP_899051.1 | |
| NM_183229.2 | 7 | 4930 | NP_899052.1 | |
| NM_183230.2 | 7 | 4930 | NP_899053.1 | |
| NM_183231.2 | 6 | 4762 | NP_899054.1 | |
| NM_183232.2 | 6 | 4813 | NP_899055.1 |
More Information
Set Membership: |
|

Genomic Map