Assay Details
Target Gene Details
Entrez Gene ID: | 23533 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 5 |
Gene Aliases: |
F730038I15Rik, FOAP-2, P101-PI3K, p101 |
Location: |
Chr.17:8878916-8965712 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3R5 | NM_001142633.2 | 19 | 4070 | NP_001136105.1 |
| NM_001251851.1 | 19 | 4132 | NP_001238780.1 | |
| NM_001251852.1 | 18 | 3954 | NP_001238781.1 | |
| NM_001251853.1 | 18 | 3916 | NP_001238782.1 | |
| NM_001251855.1 | 18 | 3895 | NP_001238784.1 | |
| NM_014308.3 | 19 | 4007 | NP_055123.2 | |
| XM_005256580.3 | 19 | 4062 | XP_005256637.1 | |
| BC028212.1 | 18 | 3886 | AAH28212.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1970 | Chr.17:8843174 - 8888125 on Build GRCh38 | Deletion |
|
| nsv510696 | Chr.17:8872217 - 8929227 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map