Assay Details
Target Gene Details
Entrez Gene ID: | 54828 |
Gene Name: | BCAS3, microtubule associated cell migration factor |
Gene Aliases: |
GAOB1, MAAB |
Location: |
Chr.17:60677811-61392838 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061134 | Chr.17:60647092 - 60876233 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map