Assay Details
Target Gene Details
Entrez Gene ID: | 92521 |
Gene Name: | sperm antigen with calponin homology and coiled-coil domains 1 |
Gene Aliases: |
CYTSB, HCMOGT-1, HCMOGT1, NSP |
Location: |
Chr.17:20009301-20319026 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPECC1 | NM_001243439.1 | NP_001230368.1 | ||
| XM_017025319.1 | XP_016880808.1 | |||
| XM_017025320.1 | XP_016880809.1 | |||
| XM_017025325.1 | XP_016880814.1 | |||
| AK295093.1 | ||||
| DA058164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3572476 | Chr.17:20044608 - 20342924 on Build GRCh38 | Gain |
|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
| nsv833394 | Chr.17:19924401 - 20096264 on Build GRCh38 | Gain |
|

Genomic Map