Assay Details
Target Gene Details
Entrez Gene ID: | 51649 |
Gene Name: | mitochondrial ribosomal protein S23 |
Gene Aliases: |
CGI-138, HSPC329, MRP-S23 |
Location: |
Chr.17:57838926-57850072 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS23 | NM_016070.3 | NP_057154.2 | ||
| AF151896.1 | AAD34133.1 | |||
| AF161447.1 | AAF29007.1 | |||
| AK312729.1 | ||||
| BC000242.1 | AAH00242.1 | |||
| BX479815.1 |
More Information
Set Membership: |
|

Genomic Map